Sodium flavonol-2'-sulphonate hydrate ≥98.0% (by titrimetric analysis) for determination of Sn, Zr
Catalog # TCIAF0184-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Ion exchange titration):min. 98.0% (calcd.on anh.substance)
- Water:max. 8.0 %
Specification Test Results
About this item
:
:340.28 g/mol
CAS Number:77125-87-2
Formula:C₁₅H₁₀O₆S
MDL Number:MFCD00059757