Fast red ITR base ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF0089-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:103.0 to 107.0 deg-C
Specification Test Results
About this item
:
:258.34 g/mol
EINECS:202-575-5
CAS Number:97-35-8
Formula:C₁₁H₁₈N₂O₃S
MDL Number:MFCD00009045