
(Ethyl)triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:896-33-3
:
Formula:C₂₀H₂₀ClP
MDL Number:MFCD00044699
:326.81 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:241.0 to 245.0 deg-C
Specification Test Results
Frequently Bought Together