
Ethylene glycolbismercaptoacetate ≥97.0% (by GC, titration analysis), purified
TCIAE1333-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Iodometric Titration):min. 97.0 %
:
:210.28 g/mol
:139°C
:GHS07
:123-81-9
:C₆H₁₀O₄S₂
:MFCD00004875