Order Entry
Netherlands
ContactUsLinkComponent
 
Ethylhexyl triazone ≥98.0% (by total nitrogen basis)
 :  TCI
 
undefined
Ethylhexyl triazone ≥98.0% (by total nitrogen basis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    124.0 to 129.0 deg-C

 

Frequently Bought Together