
Ethyl(2-methoxyethyl)dimethylammonium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)
TCIAE1275-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Colorless to Light yellow clear liquid
- Purity (Ion exchange titration):min. 98.0 %
:
:312.35 g/mol
:GHS07
:1235234-35-1
:C₇H₁₈F₂N₂O₅S₂