
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate ≥98.0% (by GC, titration analysis)
Catalog # TCIAE1271-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
About this item
:
:194.19 g/mol
Boiling Pt:120°C
EINECS:263-421-0
:GHS07
CAS Number:62124-77-0
Formula:C₉H₁₀N₂O₃
MDL Number:MFCD04114374