
Ethylene Glycol bis(3-mercaptopropionate) ≥97.0% (by titrimetric analysis)
: TCI
:22504-50-3
:
:C₈H₁₄O₄S₂
:MFCD00053520
:238.32 g/mol
:255°C
:245-044-3
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 92.0 %
- Purity (Iodometric Titration):min. 97.0 %
Frequently Bought Together