4''-Ethyl-2'-fluoro-4-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:95759-44-7
:
Formula:C₂₃H₂₃F
MDL Number:MFCD01830216
:318.44 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:69.0 to 133.0 deg-C
Specification Test Results
Frequently Bought Together