Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:141573-95-7
:
Formula:C₈H₁₀F₂N₂O₂
MDL Number:MFCD12022341
Boiling Pt:120°C
:204.18 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
Frequently Bought Together