4-Ethyl-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:84540-37-4
:
Formula:C₂₃H₃₀
MDL Number:MFCD09751090
:306.49 g/mol
EINECS:283-130-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:134.0 to 169.0 deg-C
Specification Test Results
Frequently Bought Together