4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:189750-98-9
:MFCD23115471
Formula:C₂₃H₂₈F₂O
MDL Number:MFCD18451991
:358.47 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:77.0 to 81.0 deg-C
- Mesomorphic range:77.0 to 175.0 deg-C
Specification Test Results
Frequently Bought Together