
6-Ethoxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:352525-98-5
:
Formula:C₁₂H₁₃BO₃
MDL Number:MFCD03427009
:216.04 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
Specification Test Results
Related Information
Frequently Bought Together