Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
3-Ethoxyacryloyl chloride ≥88.0% (by GC)
  TCIAE1066-5G
 :  TCI
 :  
 
 
3-Ethoxyacryloyl chloride ≥88.0% (by GC)
  TCIAE1066-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 88.0 %
  • Purity (Argentmetric Titration):
    min. 95.0 %

 

 

 :
 :134.56 g/mol
 :75°C
 :25°C
 :Refrigerator
 :228-239-8
 :2920
 :8,II
 :GHS02
 :GHS05
 :6191-99-7
 :C₅H₇ClO₂
 :MFCD09029675