
[[(4-Ethyl-3,5-dimethyl-1H-pyrrol-2-yl)(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥98.0% (by HPLC)
Catalog # TCIAE1065-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):8.3 to 9.3 %
- Melting point:210.0 to 215.0 deg-C
- Lambda max.:518.0 to 522.0 nm (CH2Cl2)
Specification Test Results
About this item
:
:318.22 g/mol
:GHS07
CAS Number:131083-16-4
Formula:C₁₈H₂₅BF₂N₂
MDL Number:MFCD28009147