Order Entry
Netherlands
ContactUsLinkComponent
 
 
3-(2-Ethylhexyloxy)-1,2-propanediol ≥98.0% (by GC)
  TCIAE1044-25ML
 :  TCI
 :  
 
3-(2-Ethylhexyloxy)-1,2-propanediol ≥98.0% (by GC)
  TCIAE1044-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :204.31 g/mol
 :152°C
 :408-080-2
 :GHS05
 :70445-33-9
 :C₁₁H₂₄O₃
 :MFCD03790878