Ethyl-3-oxo-3-(pyridin-4-yl)propanoate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26377-17-3
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD00094021
:193.20 g/mol
Boiling Pt:169°C
EINECS:247-635-1
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:55.0 to 59.0 deg-C
Specification Test Results
Frequently Bought Together