(1R,3S,5R,7R,8aS)-7-Ethylhexahydro-1-(6-hydroxy-4-quinolinyl)-3,7-methano-1H-pyrrolo[2,1-c][1,4]oxazine ≥98.0% (by HPLC)
Catalog # TCIAE0974-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):8.5 to 9.5 %
Specification Test Results
About this item
:
:310.40 g/mol
CAS Number:1476067-44-3
Formula:C₁₉H₂₂N₂O₂