Ethyl (R)-1-(tert-butoxycarbonyl)-3-piperidinecarboxylate ≥98.0% (by GC)
Catalog # TCIAE0948-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Melting point:35.0 to 39.0 deg-C
- Specific rotation [a]20/D:-45.0 to -49.0 deg (C=0.1、CHCL3)
Specification Test Results
About this item
:
:257.33 g/mol
CAS Number:194726-40-4
Formula:C₁₃H₂₃NO₄
MDL Number:MFCD07374389