Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
4-Ethylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
  TCIAE0934-5ML
 :  TCI
 :  
 
 
4-Ethylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
  TCIAE0934-5ML
 :  TCI
 :  

 

  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :127.23 g/mol
 :80°C
 :2734
 :8,II
 :GHS02
 :GHS05
 :42195-97-1
 :C₈H₁₇N
 :MFCD04969087