
Ethyl-3,4-diaminobenzoate ≥98.0% (by GC, titration analysis)
TCIAE0930-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:110.0 to 114.0 deg-C
:
:180.21 g/mol
:GHS07
:37466-90-3
:C₉H₁₂N₂O₂
:MFCD00179337