
O-Ethylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3332-29-4
:
Formula:C₂H₈ClNO
MDL Number:MFCD00012956
:97.54 g/mol
EINECS:222-060-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:131.0 to 135.0 deg-C
Specification Test Results
Frequently Bought Together