
Ethyl-1H-tetrazole-5-acetate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13616-37-0
:
Formula:C₅H₈N₄O₂
MDL Number:MFCD00274213
:156.15 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:gt. 126.0 to 130.0 deg-C
Specification Test Results
Frequently Bought Together