Order Entry
Netherlands
ContactUsLinkComponent
1-Ethyl-3-methylimidazoliumtrifluoro(trifluoromethyl)borate ≥98.0% (by HPLC)
Irritant
1-Ethyl-3-methylimidazoliumtrifluoro(trifluoromethyl)borate ≥98.0% (by HPLC)
Catalog # TCIAE0836-5G
Supplier:  TCI
CAS Number:  
Irritant
1-Ethyl-3-methylimidazoliumtrifluoro(trifluoromethyl)borate ≥98.0% (by HPLC)
Catalog # TCIAE0836-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green clear liquid
  • Purity (HPLC):
    min. 98.0 area%
  • Water:
    max. 0.5 %

Specification Test Results

About this item

 :
 :247.98 g/mol
Melting Pt:-20°C
 :GHS07
CAS Number:681856-28-0
Formula:C₇H₁₁BF₆N₂
Melting Pt:–20°C