EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥95.0% (by titrimetric analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:67-42-5
:
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291
Merck Index:14,03529
Melting Pt:241°C
:380.35 g/mol
EINECS:200-651-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 95.0 %
- Enzyme inhibition test:to pass test
Specification Test Results
Frequently Bought Together