Ethyl-6-[4-(diphenylamino)phenyl]coumarin-3-carboxylate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1056693-13-0
:
Formula:C₃₀H₂₃NO₄
:461.52 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together