
1-(4-Ethoxyphenyl)-5-mercapto-1H-tetrazole ≥98.0% (by HPLC)
TCIAE0668-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
:
:222.27 g/mol
:160°C
:1325
:4.1,III
:GHS02
:15182-68-0
:C₉H₁₀N₄OS