
5-Ethoxy-2-mercaptobenzimidazole ≥98.0% (by HPLC)
Catalog # TCIAE0535-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
About this item
:
:194.25 g/mol
:GHS07
CAS Number:55489-15-1
Formula:C₉H₁₀N₂OS
MDL Number:MFCD01326487