
Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:381670-33-3
:
Formula:C₉H₁₇NO₂
MDL Number:MFCD00671533
:171.24 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Specific rotation [a]20/D:+90.0 to +98.0 deg (C=0.6, CHCl3)
Specification Test Results
Related Information
Frequently Bought Together