
cis-Epoxysuccinic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16533-72-5
:
Formula:C₄H₄O₅
MDL Number:MFCD00191645
:132.07 g/mol
EINECS:240-604-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:149.0 to 155.0 deg-C
Specification Test Results
Frequently Bought Together