

N-(2-Methoxyethyl)ethylamine ≥97.0% (by GC)
: TCI
:34322-82-2
:
:C₅H₁₃NO
:MFCD00144876
:103.17 g/mol
:116°C
:2734
:8,II
:GHS02
:GHS05
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together