
3-Ethoxyacrylonitrile (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:61310-53-0
:
Formula:C₅H₇NO
MDL Number:MFCD00010193
:97.12 g/mol
Boiling Pt:71°C
Flash Pt:82°C
EINECS:262-706-7
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0% (total of isomer)
Specification Test Results
Frequently Bought Together