Order Entry
Netherlands
ContactUsLinkComponent
4,4'-Ethylenedianiline ≥97.0% (by GC, titration analysis)
Irritant
Health
4,4'-Ethylenedianiline ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
Health
4,4'-Ethylenedianiline ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    134.0 to 139.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together