Order Entry
Netherlands
ContactUsLinkComponent
 
8-Ethoxyquinoline-5-sulphonic acid hydrate ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
8-Ethoxyquinoline-5-sulphonic acid hydrate ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in hot Water:
    almost transparency
  • Water:
    max. 8.0 %

 

Frequently Bought Together