Order Entry
Netherlands
ContactUsLinkComponent
Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate ≥98.0% (by titrimetric analysis)
Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAE0288-1G
Supplier:  TCI
CAS Number:  
Ethylenediamine-N,N'-diacetic-N,N'-dipropionic acid hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAE0288-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in hot Water:
    almost transparency
  • Water:
    max. 15.0 %

Specification Test Results

About this item

 :
 :320.30 g/mol
CAS Number:32701-19-2
Formula:C₁₂H₂₀N₂O₈
MDL Number:MFCD00002768