4-Ethyl-1-octyn-3-ol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5877-42-9
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00015262
:154.25 g/mol
Boiling Pt:126°C
EINECS:227-545-9
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together