Order Entry
Netherlands
ContactUsLinkComponent
N,N'-Ethylenebis(stearamide) ≥85.0% (by GC) (mixture of fatty acid amides) (consists of C14, C16 and C18)
Irritant
N,N'-Ethylenebis(stearamide) ≥85.0% (by GC) (mixture of fatty acid amides) (consists of C14, C16 and C18)
Supplier:  TCI
Certificates
Irritant
N,N'-Ethylenebis(stearamide) ≥85.0% (by GC) (mixture of fatty acid amides) (consists of C14, C16 and C18)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 85.0% (C14, C16 and C18 mixture)
  • Total Nitrogen:
    4.45 to 5.00 %
  • Melting point:
    142.0 to 147.0 deg-C

Specification Test Results

Frequently Bought Together