
Ethyltripropylammonium Iodide ≥99.0% (by titrimetric analysis)
Catalogus # TCIAE0191-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:299.24 g/mol
:GHS07
CAS Number:15066-80-5
Formula:C₁₁H₂₆IN
MDL Number:MFCD00059976