Order Entry
Netherlands
ContactUsLinkComponent
Triethylene glycol monoethyl ether ≥90.0% (by GC)
Triethylene glycol monoethyl ether ≥90.0% (by GC)
Catalog # TCIAE0167-25ML
Supplier:  TCI
CAS Number:  
Triethylene glycol monoethyl ether ≥90.0% (by GC)
Catalog # TCIAE0167-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 90.0 %

Specification Test Results

About this item

 :
 :178.23 g/mol
Boiling Pt:255°C
Melting Pt:-19°C
Melting Pt:–19°C
Flash Pt:126°C
EINECS:203-978-9
CAS Number:112-50-5
Formula:C₈H₁₈O₄
MDL Number:MFCD00020605