Order Entry
Netherlands
ContactUsLinkComponent
Ethylene sulphide ≥98.0% (by GC) stabilised
Flammable
Corrosive
Toxic
Ethylene sulphide ≥98.0% (by GC) stabilised
Catalog # TCIAE0133-10G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Toxic
Ethylene sulphide ≥98.0% (by GC) stabilised
Catalog # TCIAE0133-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :60.11 g/mol
Boiling Pt:56°C
UN:1992
ADR:3,II
 :Stabilised with butyl mercaptan
EINECS:206-993-9
 :GHS02
 :GHS05
 :GHS06
Storage Temperature:Freezer
CAS Number:420-12-2
Formula:C₂H₄S
MDL Number:MFCD00005159