
Ethylene glycolbis(propionitrile)ether ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3386-87-6
:
Formula:C₈H₁₂N₂O₂
MDL Number:MFCD00019871
:168.20 g/mol
Boiling Pt:165°C
EINECS:222-208-2
UN:3276
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together