Order Entry
Netherlands
ContactUsLinkComponent
 
Ethyl cellulose, 18-22 mPa.s [5% in Toluene + Ethanol (80:20), 25 °C]
 :  TCI
 
Ethyl cellulose, 18-22 mPa.s [5% in Toluene + Ethanol (80:20), 25 °C]
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Viscosity:
    18.0 to 22.0 mPa-s (5%, toluene+ethanol 80:20, 25deg-c)

 

Frequently Bought Together