6,6'-Dibromo-1,1'-di(n-octyl)isoindigo ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD5742-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:139.0 to 143.0 deg-C
Specification Test Results
About this item
:
:644.49 g/mol
CAS Number:1334295-79-2
Formula:C₃₂H₄₀Br₂N₂O₂