6,6'-Dibromo-1,1'-bis(2-decyltetradecyl)isoindigo ≥98.0% (by total nitrogen basis)
Catalog # TCIAD5740-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Red to Dark red to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
About this item
:
:1093.36 g/mol
CAS Number:1801592-76-6
Formula:C₆₄H₁₀₄Br₂N₂O₂