6,6'-Dibromo-1,1'-bis(2-hexyldecyl)isoindigo ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD5739-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
About this item
:
:868.92 g/mol
CAS Number:1147124-24-0
Formula:C₄₈H₇₂Br₂N₂O₂