
Dicyclohexyl-3,4,3',4'-tetracarboxylic Dianhydride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:122640-83-9
:
Formula:C₁₆H₁₈O₆
MDL Number:MFCD23134595
:306.31 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Methanolysis Method):min. 97.0 %
- Melting point:206.0 to 210.0 deg-C
Specification Test Results
Frequently Bought Together