6,7-Dihydrobenzo[b]thiophen-4-one ≥95.0% (by GC)
: TCI
:13414-95-4
:
:C₈H₈OS
:MFCD00005861
:152.211 g/mol
:92°C
:Refrigerator
:236-510-7
- Appearance:White to Light yellow to Light orange powder to lump
- Purity (GC):min. 95.0 %
Frequently Bought Together