2,7-Dibromospiro[9H-fluorene-9,9'-[9H]xanthene] ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:198142-65-3
:
Formula:C₂₅H₁₄Br₂O
MDL Number:MFCD30342413
:490.19 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together