Order Entry
Netherlands
ContactUsLinkComponent
1,3-Dibromoadamantane ≥99.0% (by GC), purified by sublimation
1,3-Dibromoadamantane ≥99.0% (by GC), purified by sublimation
Supplier:  TCI
Certificates
undefined
1,3-Dibromoadamantane ≥99.0% (by GC), purified by sublimation
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    110.0 to 114.0 deg-C

Specification Test Results

Frequently Bought Together