trans,trans-3,4-Difluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:136609-96-6
:
Formula:C₂₉H₃₈F₂
MDL Number:MFCD11053474
:424.62 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:95.0 to 263.0 deg-C
Specification Test Results
Frequently Bought Together